Assay Details
Gene Symbol: MINDY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498178.6 | XP_001498228.1 | 887 | 73 | ||
XM_023632239.1 | XP_023488007.1 | 399 | 73 | |||
XM_023632240.1 | XP_023488008.1 | 299 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
