Assay Details
Gene Symbol: FBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632011.1 | XP_023487779.1 | 2012 | 64 | ||
XM_023632012.1 | XP_023487780.1 | 1858 | 64 | |||
XM_023632013.1 | XP_023487781.1 | 2012 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
