Assay Details
Gene Symbol: CNIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654494.1 | XP_023510262.1 | 538 | 92 | ||
XM_023654495.1 | XP_023510263.1 | 389 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
