Assay Details
Gene Symbol: ZNF384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610874.3 | XP_005610931.1 | 267 | 75 | ||
XM_005610875.3 | XP_005610932.1 | 815 | 75 | |||
XM_005610876.3 | XP_005610933.1 | 816 | 75 | |||
XM_005610877.3 | XP_005610934.1 | 813 | 75 | |||
XM_014740608.2 | XP_014596094.1 | 267 | 75 | |||
XM_014740610.2 | XP_014596096.1 | 325 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
