Assay Details
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619427.1 | XP_023475195.1 | 257 | 87 | ||
XM_023619429.1 | XP_023475197.1 | 262 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
