Assay Details
Gene Symbol: ELK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739454.2 | XP_014594940.1 | 1664 | 62 | ||
XM_023640412.1 | XP_023496180.1 | 1429 | 62 | |||
GenBank mRNA | JL623167.1 | - | 2 - 3 | 845 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
