Assay Details
Gene Symbol: SNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488056.5 | XP_001488106.1 | 259 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
