Assay Details
Gene Symbol: SMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637175.1 | XP_023492943.1 | 2263 | 53 | ||
XM_023637176.1 | XP_023492944.1 | 2196 | 53 | |||
XM_023637177.1 | XP_023492945.1 | 2079 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
