Assay Details
Gene Symbol: MNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653231.1 | XP_023508999.1 | 1419 | 69 | ||
XM_023653232.1 | XP_023509000.1 | 1137 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
