Assay Details
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639500.1 | XP_023495268.1 | 1277 | 75 | ||
XM_023639506.1 | XP_023495274.1 | 1183 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
