Assay Details
Gene Symbol: MAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645691.1 | XP_023501459.1 | 2096 | 70 | ||
XM_023645692.1 | XP_023501460.1 | 2097 | 70 | |||
XM_023645693.1 | XP_023501461.1 | 2095 | 70 | |||
XM_023645694.1 | XP_023501462.1 | 2095 | 70 | |||
XM_023645695.1 | XP_023501463.1 | 2094 | 70 | |||
XM_023645696.1 | XP_023501464.1 | 2096 | 70 | |||
XM_023645697.1 | XP_023501465.1 | 2094 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
