Assay Details
Gene Symbol: STARD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917447.5 | XP_001917482.1 | 381 | 79 | ||
XM_023645808.1 | XP_023501576.1 | 465 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
