Assay Details
Gene Symbol: RRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001918081.4 | XP_001918116.2 | 1046 | 59 | ||
XM_023645906.1 | XP_023501674.1 | 1046 | 59 | |||
XM_023645907.1 | XP_023501675.1 | 1046 | 59 | |||
XM_023645908.1 | XP_023501676.1 | 1046 | 59 | |||
XM_023645910.1 | XP_023501678.1 | 1046 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
