Assay Details
Gene Symbol: LOC106783453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645913.1 | XP_023501681.1 | 684 | 102 | ||
XM_023645915.1 | XP_023501683.1 | 3815 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023645913.1, XM_023645915.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
