Assay Details
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645972.1 | XP_023501740.1 | 702 | 84 | ||
XM_023645973.1 | XP_023501741.1 | 702 | 84 | |||
XM_023645974.1 | XP_023501742.1 | 702 | 84 | |||
XM_023645976.1 | XP_023501744.1 | 2877 | 84 | |||
XM_023645978.1 | XP_023501746.1 | 2769 | 84 | |||
XM_023645979.1 | XP_023501747.1 | 936 | 84 | |||
XM_023645980.1 | XP_023501748.1 | 1545 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
