Assay Details
Gene Symbol: DENND2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504905.5 | XP_001504955.1 | 2712 | 60 | ||
XM_005612148.3 | XP_005612205.1 | 2533 | 60 | |||
XM_005612149.3 | XP_005612206.1 | 2630 | 60 | |||
XM_014741799.2 | XP_014597285.1 | 2774 | 60 | |||
XM_023646005.1 | XP_023501773.1 | 2863 | 60 | |||
XM_023646006.1 | XP_023501774.1 | 1444 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
