Assay Details
Gene Symbol: LOC102147743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621809.1 | XP_023477577.1 | 1660 | 103 | ||
XM_023621810.1 | XP_023477578.1 | 1799 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
