Assay Details
Gene Symbol: LOC102147743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621809.1 | XP_023477577.1 | 1250 | 110 | ||
XM_023621810.1 | XP_023477578.1 | 1389 | 110 | |||
GenBank mRNA | JL640738.1 | - | 4 - 5 | 794 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
