Assay Details
Gene Symbol: NAXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601339.2 | XP_005601396.1 | 569 | 57 | ||
XM_005601340.3 | XP_005601397.1 | 591 | 57 | |||
XM_023621836.1 | XP_023477604.1 | 569 | 57 | |||
XM_023621837.1 | XP_023477605.1 | 694 | 57 | |||
XM_023621838.1 | XP_023477606.1 | 681 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
