Assay Details
Gene Symbol: PROC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023622666.1 | XP_023478434.1 | 387 | 81 | ||
XM_023622667.1 | XP_023478435.1 | 448 | 81 | |||
XM_023622668.1 | XP_023478436.1 | 384 | 81 | |||
XM_023622669.1 | XP_023478437.1 | 445 | 81 | |||
XM_023622670.1 | XP_023478438.1 | 398 | 81 | |||
XM_023622671.1 | XP_023478439.1 | 318 | 81 | |||
XM_023622673.1 | XP_023478441.1 | 303 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
