Assay Details
Gene Symbol: DBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001487877.4 | XP_001487927.1 | 323 | 95 | ||
XM_023622784.1 | XP_023478552.1 | 317 | 95 | |||
XM_023622785.1 | XP_023478553.1 | 475 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
