Assay Details
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488904.4 | XP_001488954.2 | 905 | 87 | ||
XM_023622811.1 | XP_023478579.1 | 1076 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
