Assay Details
Gene Symbol: CCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915413.5 | XP_001915448.1 | 577 | 79 | ||
XM_005601461.3 | XP_005601518.1 | 577 | 79 | |||
XM_014732439.2 | XP_014587925.1 | 717 | 79 | |||
XM_014732440.2 | XP_014587926.1 | 558 | 79 | |||
XM_014732441.2 | XP_014587927.1 | 196 | 79 | |||
XM_023622827.1 | XP_023478595.1 | 686 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
