Assay Details
Gene Symbol: PPIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617796.1 | XP_023473564.1 | 391 | 67 | ||
XM_023617797.1 | XP_023473565.1 | 400 | 67 | |||
XM_023617798.1 | XP_023473566.1 | 391 | 67 | |||
XM_023617799.1 | XP_023473567.1 | 391 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
