Assay Details
Gene Symbol: GLRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504609.6 | XP_001504659.1 | 312 | 83 | ||
XM_005599547.3 | XP_005599604.1 | 313 | 83 | |||
GenBank mRNA | JL621136.1 | - | 1 - 2 | 316 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
