Assay Details
Gene Symbol: CCNH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504626.5 | XP_001504676.1 | 329 | 91 | ||
XM_023618036.1 | XP_023473804.1 | 332 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
