Assay Details
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730791.2 | XP_014586277.1 | 805 | 69 | ||
XM_023618052.1 | XP_023473820.1 | 970 | 69 | |||
XM_023618053.1 | XP_023473821.1 | 2797 | 69 | |||
XM_023618054.1 | XP_023473822.1 | 953 | 69 | |||
XM_023618055.1 | XP_023473823.1 | 1207 | 69 | |||
XM_023618056.1 | XP_023473824.1 | 862 | 69 | |||
XM_023618057.1 | XP_023473825.1 | 1057 | 69 | |||
XM_023618058.1 | XP_023473826.1 | 1000 | 69 | |||
GenBank mRNA | JL616466.1 | - | 7 - 8 | 693 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
