Assay Details
Gene Symbol: STK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493184.6 | XP_001493234.1 | 475 | 70 | ||
XM_014740380.2 | XP_014595866.1 | 463 | 70 | |||
XM_023642387.1 | XP_023498155.1 | 475 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
