Assay Details
Gene Symbol: SGPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493041.4 | XP_001493091.1 | 836 | 72 | ||
XM_023642421.1 | XP_023498189.1 | 679 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
