Assay Details
Gene Symbol: CCL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365131.4 | XP_003365179.2 | 1402 | 107 | ||
XM_005610663.3 | XP_005610720.1 | 1388 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
