Assay Details
Gene Symbol: HIBADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499874.5 | XP_001499924.1 | 596 | 83 | ||
XM_014739146.1 | XP_014594632.1 | 600 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
