Assay Details
Gene Symbol: ZNRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639452.1 | XP_023495220.1 | 700 | 113 | ||
XM_023639453.1 | XP_023495221.1 | 700 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
