Assay Details
Gene Symbol: PPP1R17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499697.5 | XP_001499747.3 | 197 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001499697.5 transcripts.
Set Membership: |
![]() ![]() |
