Assay Details
Gene Symbol: NT5C3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500985.6 | XP_001501035.3 | 640 | 80 | ||
XM_005609287.3 | XP_005609344.1 | 945 | 80 | |||
XM_005609288.3 | XP_005609345.1 | 814 | 80 | |||
XM_005609289.3 | XP_005609346.1 | 705 | 80 | |||
XM_005609290.3 | XP_005609347.1 | 876 | 80 | |||
GenBank mRNA | JL641549.1 | - | 1 - 2 | 170 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
