Assay Details
Gene Symbol: TSPAN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502043.6 | XP_001502093.1 | 640 | 67 | ||
XM_014739138.2 | XP_014594624.1 | 437 | 67 | |||
XM_023639555.1 | XP_023495323.1 | 693 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
