Assay Details
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501290.4 | XP_001501340.1 | 1606 | 76 | ||
XM_014739205.2 | XP_014594691.1 | 1697 | 76 | |||
XM_023639580.1 | XP_023495348.1 | 1756 | 76 | |||
XM_023639581.1 | XP_023495349.1 | 1611 | 76 | |||
XM_023639582.1 | XP_023495350.1 | 1606 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
