Assay Details
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502549.6 | XP_001502599.3 | 367 | 59 | ||
XM_023639620.1 | XP_023495388.1 | 306 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
