Assay Details
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618330.1 | XP_023474098.1 | 796 | 64 | ||
XM_023618331.1 | XP_023474099.1 | 704 | 64 | |||
XM_023618332.1 | XP_023474100.1 | 625 | 64 | |||
XM_023618333.1 | XP_023474101.1 | 572 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
