Assay Details
Gene Symbol: MRPL55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730843.2 | XP_014586329.1 | 143 | 115 | ||
XM_014730844.2 | XP_014586330.1 | 131 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014730843.2, XM_014730844.2 transcripts.
Set Membership: |
![]() ![]() |
