Assay Details
Gene Symbol: MRPL55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730842.2 | XP_014586328.1 | 442 | 85 | ||
XM_014730843.2 | XP_014586329.1 | 430 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
