Assay Details
Gene Symbol: ARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618336.1 | XP_023474104.1 | 573 | 106 | ||
XM_023618337.1 | XP_023474105.1 | 443 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
