Assay Details
Gene Symbol: SH3RF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618447.1 | XP_023474215.1 | 3067 | 52 | ||
XM_023618448.1 | XP_023474216.1 | 2971 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() |
