Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488049.6 | XP_001488099.1 | 347 | 99 | ||
XM_023618487.1 | XP_023474255.1 | 392 | 99 | |||
XM_023618488.1 | XP_023474256.1 | 208 | 99 | |||
XM_023618489.1 | XP_023474257.1 | 209 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001488049.6, XM_023618487.1, XM_023618488.1, XM_023618489.1 transcripts.
Set Membership: |
![]() ![]() |
