Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488049.6 | XP_001488099.1 | 1311 | 77 | ||
XM_005599781.3 | XP_005599838.1 | 1310 | 77 | |||
XM_023618487.1 | XP_023474255.1 | 1356 | 77 | |||
XM_023618488.1 | XP_023474256.1 | 1172 | 77 | |||
XM_023618489.1 | XP_023474257.1 | 1173 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
