Assay Details
Gene Symbol: TMEM182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488300.4 | XP_001488350.1 | 402 | 87 | ||
XM_023618504.1 | XP_023474272.1 | 404 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
