Assay Details
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618498.1 | XP_023474266.1 | 792 | 93 | ||
XM_023618499.1 | XP_023474267.1 | 737 | 93 | |||
XM_023618500.1 | XP_023474268.1 | 711 | 93 | |||
XM_023618501.1 | XP_023474269.1 | 1118 | 93 | |||
XM_023618502.1 | XP_023474270.1 | 796 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
