Assay Details
Gene Symbol: MRPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599799.3 | XP_005599856.1 | 502 | 79 | ||
XM_023618580.1 | XP_023474348.1 | 486 | 79 | |||
XM_023618581.1 | XP_023474349.1 | 514 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
