Assay Details
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492041.5 | XP_001492091.1 | 798 | 67 | ||
XM_003362917.4 | XP_003362965.1 | 831 | 67 | |||
XM_023618654.1 | XP_023474422.1 | 778 | 67 | |||
XM_023618655.1 | XP_023474423.1 | 1369 | 67 | |||
XM_023618656.1 | XP_023474424.1 | 721 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
