Assay Details
Gene Symbol: RTKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500407.5 | XP_001500457.2 | 1530 | 73 | ||
XM_003362966.4 | XP_003363014.3 | 1774 | 73 | |||
XM_014731082.2 | XP_014586568.2 | 1750 | 73 | |||
XM_023618839.1 | XP_023474607.1 | 1570 | 73 | |||
XM_023618840.1 | XP_023474608.1 | 1780 | 73 | |||
XM_023618841.1 | XP_023474609.1 | 1111 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
