Assay Details
Gene Symbol: MTHFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500723.3 | XP_001500773.1 | 935 | 78 | ||
XM_005599899.3 | XP_005599956.1 | 916 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
